Boron nitride, low binder, 99.5% (metals basis), Alfa Aesar™
Used as a lubricant and an additive to cosmetic products. Boron nitride has potential use in nanotechnology. Also used in paints, dental cements, pencil leads, xerographic process and laser printers, sealing oxygen sensors.
Manufacturer:  Alfa Aesar 45573KJ
 View more versions of this product
Catalog No. AA45573KJ
Specifications
Specifications
Boron nitride
BN
boron nitride, elbor, boron nitride, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
B#N
24.817
CHEBI:50883
99.5%
5 pc.
10043-11-5
MFCD00011317
PZNSFCLAULLKQX-UHFFFAOYSA-N
azanylidyneborane
66227
24.82
Bag
Chemical Identifiers
10043-11-5
24.817
PZNSFCLAULLKQX-UHFFFAOYSA-N
66227
azanylidyneborane
BN
MFCD00011317
boron nitride, elbor, boron nitride, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
CHEBI:50883
B#N
Safety and Handling
Safety and Handling
TSCA : Yes
SDS Documents