missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron nitride, Liquid, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4408618

 View more versions of this product

Catalog No. AA4408618



Specifications

Specifications

Boron nitride
BN
borazon
PZNSFCLAULLKQX-UHFFFAOYSA-N
azanylidyneborane
66227
Paste
10043-11-5
MFCD00011317
Soluble in water (slightly soluble) at 20°C,and water (soluble) at 95°C.
B#N
24.817
CHEBI:50883
50g

Chemical Identifiers

10043-11-5
24.817
PZNSFCLAULLKQX-UHFFFAOYSA-N
66227
azanylidyneborane
BN
MFCD00011317
borazon
CHEBI:50883
B#N
Safety and Handling

Safety and Handling

H301-H332: Toxic if swallowed. Harmful if inhaled.

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents