Learn More
Boron nitride, Refractory Brushable Paint, BN 31%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04408630
Description
Boron nitride is used as a lubricant and an additive to cosmetic products. It is also used in parts of high-temperature equipment, nanotechnology, polymorph. It is also used in used in xerographic process and laser printers. It is also used in fiberglass, ceramics, wood preservative, nuclear application and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boron nitride | |
Paste | |
BN | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
azanylidyneborane | |
66227 |
10043-11-5 | |
250 g | |
MFCD00011317 | |
Soluble in water (slightly soluble) at 20°C,and water (soluble) at 95°C. | |
B=N | |
24.82 | |
CHEBI:50883 |
Chemical Identifiers
10043-11-5 | |
24.82 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
66227 | |
B=N |
BN | |
MFCD00011317 | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
CHEBI:50883 |
Safety and Handling
H301-H332: Toxic if swallowed. Harmful if inhaled.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only