missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron nitride, 99.5% (metals basis), Thermo Scientific Chemicals
$365.42 - $365.42
Chemical Identifiers
| CAS | 10043-11-5 |
|---|---|
| Molecular Formula | BN |
| Molecular Weight (g/mol) | 24.82 |
| MDL Number | MFCD00011317 |
| InChI Key | LNLSXDSWJBUPHM-UHFFFAOYSA-N |
| Synonym | boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r |
| PubChem CID | 66227 |
| ChEBI | CHEBI:50883 |
| SMILES | B=N |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10043-11-5 | |
| 24.82 | |
| LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
| 66227 | |
| B=N |
| BN | |
| MFCD00011317 | |
| boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
| CHEBI:50883 |
Specifications
| 10043-11-5 | |
| 1 Pc. | |
| MFCD00011317 | |
| LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
| azanylidyneborane | |
| 66227 | |
| 24.82 | |
| Bag |
| Hot Pressed Rod | |
| BN | |
| boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
| B=N | |
| 24.82 | |
| CHEBI:50883 | |
| 99.5% | |
| Boron nitride |
Safety and Handling
P261-P271-P304+P340-P312-P501c
H335
EINECSNumber : 233-136-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only