missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron nitride, 99.5% (metals basis), Thermo Scientific™
Used in production of alloys. It is also used in the manufacture of furnace refractories, insulation materials, machinery and aviation.
$838.52 - $838.52
Chemical Identifiers
| CAS | 10043-11-5 |
|---|---|
| Molecular Formula | BN |
| Molecular Weight (g/mol) | 24.82 |
| MDL Number | MFCD00011317 |
| InChI Key | LNLSXDSWJBUPHM-UHFFFAOYSA-N |
| Synonym | boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r |
| PubChem CID | 66227 |
| ChEBI | CHEBI:50883 |
| IUPAC Name | boranylidyneamine |
| SMILES | B=N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 10043-11-5 | |
| 24.82 | |
| LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
| 66227 | |
| boranylidyneamine |
| BN | |
| MFCD00011317 | |
| boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
| CHEBI:50883 | |
| B=N |
Specifications
| 10043-11-5 | |
| 1 Pc. | |
| MFCD00011317 | |
| Insoluble in water. | |
| B=N | |
| 24.82 | |
| CHEBI:50883 | |
| 99.5% | |
| Boron nitride |
| Hot Pressed Rod | |
| BN | |
| boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
| LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
| boranylidyneamine | |
| 66227 | |
| 24.82 | |
| Bag |
Safety and Handling
P261-P271-P304+P340-P312-P501c
H335
EINECSNumber : 233-136-6
RTECSNumber : ED7800000
TSCA : Yes
Recommended Storage : Ambient temperatures