missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron nitride, 99.5% (metals basis), Alfa Aesar™

Used in production of alloys. It is also used in the manufacture of furnace refractories, insulation materials, machinery and aviation.

Manufacturer:  Alfa Aesar 44838KJ

 View more versions of this product

Catalog No. AA44838KJ



Specifications

Specifications

Boron nitride
5 pc.
BN
boron nitride, elbor, boron nitride, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
B#N
24.817
CHEBI:50883
99.5%
Bag
10043-11-5
MFCD00011317
PZNSFCLAULLKQX-UHFFFAOYSA-N
azanylidyneborane
66227
24.82

Chemical Identifiers

10043-11-5
24.817
PZNSFCLAULLKQX-UHFFFAOYSA-N
66227
azanylidyneborane
BN
MFCD00011317
boron nitride, elbor, boron nitride, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
CHEBI:50883
B#N
Safety and Handling

Safety and Handling

TSCA : Yes

SDS Documents