Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron nitride, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Boron nitride
CAS 10043-11-5
Name Note Hot pressed rod, 9.5mm dia x 305mm long
Formula Weight 24.82g/mol
MDL Number MFCD00011317
Merck Index 14,1346
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40461KG

 
alfa aesar™
40461-KG
1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40461KJ

 
alfa aesar™
40461-KJ
5 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
Quantity 1 piece
Quantity 1 piece
Chemical Name or Material Boron nitride
Name Note Hot pressed rod, 9.5mm dia x 305mm long
CAS 10043-11-5
Physical Form Hot pressed rod
Assay 99.5%
Assay Percent Notes (metals basis)
Formula Weight 24.82g/mol
MDL Number MFCD00011317
Merck Index 14,1346
TSCA TSCA