Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron nitride

Chemical Identifiers

Chemical Name or Material Boron nitride
CAS 10043-11-5
Formula Weight 24.82
MDL Number MFCD00011317
Merck Index 14,1346
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4060804

  SDS
alfa aesar™
4060804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4060809

  SDS
alfa aesar™
4060809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 24.82
Chemical Name or Material Boron nitride
Health Hazard 2 GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00011317
CAS 10043-11-5
Physical Form Powder
TSCA TSCA
Merck Index 14,1346
Health Hazard 1 Warning
Mesh Size −120 Mesh
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 2g
Odor Odorless
Density 3.48g/mL
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.