Please login to your online account to display your discounted pricing

Alfa Aesar™ Boron carbide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis)

Magnetron sputtering source

Chemical Identifiers

Chemical Name or Material Boron carbide sputtering target
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Formula Weight 55.26
MDL Number MFCD00011520
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41048KS

 
alfa aesar™
41048KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 55.26
TSCA TSCA
Assay Percent Notes (metals basis)
Chemical Name or Material Boron carbide sputtering target
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.5%
MDL Number MFCD00011520
Quantity 1 each