missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boric Anhydride (Laboratory), Fisher Chemical

$226.50 - $1,240.79

Chemical Identifiers

CAS 1303-86-2
Physical Form Solid
Assay Percent Range ≥98 %
Molecular Formula B2O3
Formula Weight 69.62
MDL Number MFCD00011315
Synonym Boron Trioxide
Chemical Name or Material Boric Anhydride
Assay ≥98%
Grade Laboratory Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
A76-3 SDS Fisher Scientific Company
FLA763
Glass Bottle 3kg Each for $1,240.79
Case of 4 Each for $4,963.16
A76-500 SDS Fisher Scientific Company
FLA76500
Glass Bottle 500g Each for $226.50
Case of 6 Each for $1,359.00
Specifications Description & Specifications

Chemical Identifiers

1303-86-2
≥98 %
69.62
Boron Trioxide
≥98%
Solid
B2O3
MFCD00011315
Boric Anhydride
Laboratory Grade

Specifications

1303-86-2
≥98 %
69.62
Boron Trioxide
Pass Test
1860°C
450°C
3kg
≥98%
Solid
B2O3
MFCD00011315
2.460g/cm³
Glass Bottle
White
4
Boric Anhydride
Laboratory Grade

Safety and Handling

WARNING!

Emergency Overview
May impair fertility. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:1