missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Powder, ACS, 99.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor B112545KGBL
Specifications
| 10043-35-3 | |
| 1e-05 | |
| 45 kg | |
| MFCD00011337 | |
| OB(O)O | |
| 61.83 | |
| ACS |
| 1 | |
| Fiber Drum | |
| BH3O3 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| boric acid | |
| 99.5% |
Chemical Identifiers
| 10043-35-3 | |
| 61.83 | |
| KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
| OB(O)O |
| BH3O3 | |
| MFCD00011337 | |
| boric acid |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.