missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boric Acid, Certified, 2.0% (w/v) ±0.1%, LabChem™

$50.06 - $100.09

Chemical Identifiers

CAS 7732-18-5,10043-35-3
CAS Max % 98,2
Linear Formula H3BO3
Formula Weight 61.83
Chemical Name or Material Boric Acid
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC117204 Lab Chem
LC117204
4L Each for $100.09
LC117202 Lab Chem
LC117202
1L Each for $50.06
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,10043-35-3
H3BO3
Boric Acid
98,2
61.83
Certified Grade

Specifications

61.83
Liquid
Poly Bottle
98,2
H3BO3
Clear, colorless
Room Temperature
2.0% ± 0.1% (w/v)
Traceable to NIST
10043-35-3,7732-18-5
Passes Test
Boron
Soluble in water
4L
Boric Acid
Certified Grade

Safety and Handling

GHS H Statement
May damage fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger