Please login to your online account to display your discounted pricing

Boric Acid, ACS Grade, ≥ 99.5%, LabChem™

$69.57 - $69.57

Chemical Identifiers

Chemical Name or Material Boric Acid
CAS 10043-35-3
CAS Max % 100
Purity Grade ACS Grade
Linear Formula H3BO3
Formula Weight 61.83
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC117151

 
lab chem
LC117151
500g Each for $69.57
Description & Specifications

Specifications

Formula Weight 61.83
Assay ≥99.5%
Chemical Name or Material Boric Acid
Specific Gravity 1.435g/cm3
Color White
Storage Information Room Temperature
Health Hazard 1 Danger
Purity Grade ACS Grade
Identification Passes Test
Packaging Poly Bottle
EINECS Number 233-139-2
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
Solubility Soluble in water
Melting Point 169°C
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Quantity 500g
Linear Formula H3BO3
CAS Max % 100
Density 1.435g/cm3
Boiling Point 300°C
CAS 10043-35-3
Decomposition Information Fume; Boron
Physical Form Powder
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May damage fertility or the unborn child.