missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boric acid, 99.8%, Alfa Aesar

Manufacturer:  Alfa Aesar 12680A1

 View more versions of this product

Catalog No. AA12680A1



Specifications

Specifications

Boric acid
1kg
White granular
BH3O3
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
B(O)(O)O
1.435g/cm3
7628
61.83
Odorless
White
10043-35-3
99.8%
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
61.831
CHEBI:33118
Granular

Chemical Identifiers

10043-35-3
61.831
KGBXLFKZBHKPEV-UHFFFAOYSA-N
7628
boric acid
BH3O3
MFCD00011337
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
CHEBI:33118
B(O)(O)O
Safety and Handling

Safety and Handling

H360-H303

EINECSNumber : 233-139-2

storageNote1 : Hygroscopic

Documents