missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boric Acid, 4% (w/v), with Bromocresol Green-Methyl Red Indicator, Ricca Chemical
For Analysis of Nitrogen (Kjeldahl)
Supplier: Ricca Chemical Company 107025
Specifications
Boric Acid | |
With Bromocresol Green-Methyl Red Indicator | |
3.77,94.0,9.0E-4,7.0E-4 | |
3.92,98.0,9.0E-4,7.0E-4 | |
Aqueous Solution | |
BH3O3 | |
MFCD00011337 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
boric acid | |
7628 |
Orange-Red | |
10043-35-3,7732-18-5,62625-32-5,845-10-3 | |
4% (w/v) | |
Cubitainer | |
10 L | |
Liquid | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe | |
OB(O)O | |
61.83 | |
CHEBI:33118 |
Chemical Identifiers
10043-35-3 | |
61.83 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
7628 | |
boric acid |
BH3O3 | |
MFCD00011337 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe | |
CHEBI:33118 | |
OB(O)O |