Please login to your online account to display your discounted pricing

Boric Acid, 4% (w/v) Aq Soln, Ricca Chemical


Quantity 4L

Manufacturer: ricca chemical company  RI10681

 View more versions of this product

Catalog No. 1068-1

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 4L
Chemical Name or Material Boric Acid Solution
Name Note 4%(w/v) Aqueous Solution
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 98.0
CAS 10043-35-3
CAS Min % 3.77
CAS Max % 3.93

Specifications

Chemical Name or Material Boric Acid Solution
Name Note 4%(w/v) Aqueous Solution
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 98.0
CAS 10043-35-3
CAS Min % 3.77
CAS Max % 3.93