Please login to your online account to display your discounted pricing

Boric Acid, 2% (w/v) with Indicator (Methylene Blue-Methyl Red) for Nitrogen (Ammonia), Nitrogen (Organic), Certified, LabChem™

$63.86 - $207.17

Chemical Identifiers

Chemical Name or Material Boric Acid
CAS 7732-18-5
CAS Max % 96
CAS 10043-35-3
CAS Max % 2
CAS 64-17-5
CAS Max % 1
CAS 64-17-5
CAS Max % 0.9
CAS 67-63-0
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC117701

 
lab chem
LC117701
500mL Each for $63.86

LC117702

 
lab chem
LC117702
1L Each for $88.80

LC117704

 
lab chem
LC117704
4L Each for $207.17
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 61.83
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS 67-63-0
Health Hazard 1 Danger
CAS Max % 0.052
Physical Form Liquid
Identification Passes Test
Recommended Storage Room Temperature
CAS Max % 2
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
Solubility Soluble in water
CAS 64-17-5
Color Clear purple
CAS 7732-18-5
Quantity 500mL
CAS Max % 0.9
Name Note 2% (w/v) with Indicator (Methylene Blue-Methyl Red) for Nitrogen (Ammonia), Nitrogen (Organic)
Chemical Name or Material Boric Acid
CAS Max % 96
Linear Formula H3BO3
CAS Max % 1
Suitability Passes Test
CAS 64-17-5
CAS 10043-35-3
Decomposition Information Boron; Carbon monoxide; Carbon dioxide
Packaging Poly Bottle
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May damage fertility or the unborn child.

Room Temperature