missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boric Acid, 2% (w/v) Aqueous Solution, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI10655

 View more versions of this product

Catalog No. 10655


 

Specifications Description & Specifications

Specifications

Liquid
20L
Aqueous Solution
Cubitainer
1.92,96.0
Clear colorless
2% (w/v)
7732-18-5,10043-35-3
2.0,100.0
Boric Acid

Chemical Identifiers

7732-18-5,10043-35-3
100.0,2.0
Boric Acid
96.0,1.92
Liquid