missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Borax Standard Buffer, Fisher Chemical

Chemical Identifiers

CAS 7732-18-5,1303-96-4
CAS Min % 98.0,0.37
CAS Max % 100.0,0.39
Chemical Name or Material Borax Standard Buffer
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
SB9200-1 Ricca Chemical Company
SB92001
Poly Bottle 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,1303-96-4
98.0,0.37
100.0,0.39
Borax Standard Buffer

Specifications

1L
Poly Bottle
100.0,0.39
Borax Standard Buffer
98.0,0.37
For AOAC Method 946.01
7732-18-5,1303-96-4

Safety and Handling