Please login to your online account to display your discounted pricing

Borax, For Silica, 1% (w/v), LabChem™

$45.94 - $45.94

Chemical Identifiers

Chemical Name or Material Borax
CAS 7732-18-5
CAS Max % 99
CAS 1303-96-4
CAS Max % 1
Name Note For Silica
Linear Formula Na2B4O7·10H2O
Formula Weight 381.37
Synonym Sodium Borate; Sodium Tetraborate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC117101

 
lab chem
LC117101
500mL Each for $45.94
Description & Specifications

Specifications

Formula Weight 381.37
CAS 7732-18-5
Health Hazard 1 Danger
Packaging Poly Bottle
Assay 1% (w/v)
Identification Passes Test
Name Note For Silica
Color Clear, colorless
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
Specific Gravity 1g/mL
CAS Max % 1
Chemical Name or Material Borax
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Physical Form Liquid
Synonym Sodium Borate; Sodium Tetraborate
Linear Formula Na2B4O7·10H2O
Storage Information Room Temperature
CAS Max % 99
Solubility Soluble in water
Decomposition Information Boron
Density 1g/mL
CAS 1303-96-4
Quantity 500mL
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May damage fertility or the unborn child.