Please login to your online account to display your discounted pricing

Alfa Aesar™ Borate-buffered saline (5X, pH 7.4), low endotoxin

Chemical Identifiers

Chemical Name or Material Borate-buffered saline (5X, pH 7.4)
Name Note low endotoxin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67503AE

 
alfa aesar™
J67503AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67503AK

 
alfa aesar™
J67503AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Name Note low endotoxin
TSCA TSCA
Quantity 100mL
Chemical Name or Material Borate-buffered saline (5X, pH 7.4)
Color Colorless