missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Borate-buffered saline (5X)

$74.34 - $111.18

Chemical Identifiers

CAS 1330-43-4
MDL Number MFCD07784974
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60979AK View Documents Alfa Aesar™
J60979AK
250mL Each for $74.34
 
AAJ60979AP View Documents Alfa Aesar™
J60979AP
500mL Each for $111.18
 
Description

Description

Applications
Borate buffered saline has been used to determine whether heat-killed, borate-buffered suspensions of Gordonia bronchialis, Rhodococcus coprophilus or Tsukamurella inchonensis can be used to treat canine flea allergy. Borate buffered saline has also been used to investigate the effects of lentivirus mediated siRNA targeting human metastasis suppressor 1 (MTSS1, MIM-B gene) gene on the invasive and metastatic potentials of hepatocellular carcinoma (HCC) MHCC97H cells. Borate buffered saline has been used to determine the effects of packaging solution osmolality and buffering agent on soft contact lens parameters. Borate buffer solutions is used in protein modification procedures requiring amine-free buffer at alkaline pH.

Solubility
Soluble in water 0.1 M at 20°C.

Notes
125mM sodium borate, 750mM sodium chloride, pH 7.0
Specifications

Chemical Identifiers

1330-43-4
MFCD07784974

Specifications

1330-43-4
5X
Odorless
250mL
MFCD07784974
Liquid
Colorless
Borate-buffered saline
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA