Please login to your online account to display your discounted pricing

Borate Buffer, pH 9.5, for Ammonia and Organic Nitrogen Analysis, Ricca Chemical


For Analysis of Ammonia and organic Nitrogen

Quantity 1L
Packaging Poly Natural

Manufacturer: ricca chemical company  RI104032

 View more versions of this product

Catalog No. 1040-32

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Borate Buffer
Name Note pH 9.5
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 1303-96-4
CAS Min % 0.46
CAS Max % 0.48
CAS 1310-73-2
CAS Min % 0.07
CAS Max % 0.07
Physical Form Liquid
Packaging Poly Natural
Color Clear colorless
Quantity 1L

Specifications

Chemical Name or Material Borate Buffer
Name Note pH 9.5
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 1303-96-4
CAS Min % 0.46
CAS Max % 0.48
CAS 1310-73-2
CAS Min % 0.07
CAS Max % 0.07
Physical Form Liquid