Please login to your online account to display your discounted pricing

Alfa Aesar™ Borate, 0.5M buffer soln., pH 9.0, low endotoxin

Chemical Identifiers

Chemical Name or Material Borate
CAS 1330-43-4
Name Note low endotoxin
MDL Number MFCD00163147
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67128AE

 
alfa aesar™
J67128AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67128AK

 
alfa aesar™
J67128AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

MDL Number MFCD00163147
Chemical Name or Material Borate
pH 9.0
Color Colorless
Quantity 100mL
Concentration 0.5M buffer soln.
CAS 1330-43-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360
May damage fertility or the unborn child.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Name Note low endotoxin
Physical Form Liquid
Safety and Handling

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360
May damage fertility or the unborn child.