Please login to your online account to display your discounted pricing

Alfa Aesar™ Borate, 0.5M buffer soln., pH 9.0, low endotoxin


Quantity 250mL

Manufacturer: alfa aesar™  J67128AK

 View more versions of this product

Catalog No. AAJ67128AK

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Borate
CAS 1330-43-4
pH 9.0
Color Colorless
TSCA TSCA
Name Note low endotoxin
Physical Form Liquid
Concentration 0.5M buffer soln.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360
May damage fertility or the unborn child.
Quantity 250mL
MDL Number MFCD00163147
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Specifications

Chemical Name or Material Borate
CAS 1330-43-4
Name Note low endotoxin
MDL Number MFCD00163147
Accessories

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360
May damage fertility or the unborn child.

SDS & Safety Documents