Please login to your online account to display your discounted pricing

Alfa Aesar™ Borate, 0.5M buffer soln., pH 8.5, low endotoxin

Chemical Identifiers

Chemical Name or Material Borate
CAS 1330-43-4
Name Note low endotoxin
MDL Number MFCD00163147
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67259AE

  SDS
alfa aesar™
J67259AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67259AK

  SDS
alfa aesar™
J67259AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Borate
MDL Number MFCD00163147
pH 8.5
Color Colorless
Quantity 100mL
TSCA TSCA
Name Note low endotoxin
Physical Form Liquid
Concentration 0.5M buffer soln.
CAS 1330-43-4