Learn More
Borane-trimethylamine complex, 97%, Thermo Scientific Chemicals
$542.54 - $542.54
Chemical Identifiers
CAS | 75-22-9 |
---|---|
Molecular Formula | C3H12BN |
Molecular Weight (g/mol) | 72.95 |
MDL Number | MFCD00012420 |
InChI Key | WVMHLYQJPRXKLC-UHFFFAOYSA-N |
Synonym | Trimethylamine borane |
IUPAC Name | trimethylamine borane |
SMILES | B.CN(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC402741000
|
Thermo Scientific Chemicals
402741000 |
100 g | Glass bottle |
Each for $542.54
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
75-22-9 | |
72.95 | |
WVMHLYQJPRXKLC-UHFFFAOYSA-N | |
trimethylamine borane |
C3H12BN | |
MFCD00012420 | |
Trimethylamine borane | |
B.CN(C)C |
Specifications
75-22-9 | |
White | |
Authentic | |
Glass bottle | |
(CH3)3N·BH3 | |
100 g | |
Trimethylamine borane | |
WVMHLYQJPRXKLC-UHFFFAOYSA-N | |
trimethylamine borane | |
72.95 | |
Powder, Crystals or Chunks |
92.0°C to 96.0°C | |
172.0°C | |
97% | |
C3H12BN | |
MFCD00012420 | |
01,1229; 05,708; 12,65 | |
Solubility in water: decomposes. Other solubilities: soluble in ethanols,ethyl ether | |
B.CN(C)C | |
72.95 | |
97% | |
Borane-trimethylamine complex, 98% |
Safety and Handling
GHS H Statement:
Flammable solid.
Causes serious eye irritation.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-850-4
RTECSNumber : PA0525000
TSCA : TSCA
RUO – Research Use Only