Learn More
Borane-trimethylamine complex, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1499414
Description
Reducing agent for the preparation of plate-type palladium-zinc oxide catalys, reagent for chemical vapor generation and spectrometry determination of elements using amineboranes and cyanotrihydroborate(III) reagents. Catalyst for regioselective reductive opening of benzylidene acetal glycosides and polarity reversal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReducing agent for the preparation of plate-type palladium-zinc oxide catalys, reagent for chemical vapor generation and spectrometry determination of elements using amineboranes and cyanotrihydroborate(III) reagents. Catalyst for regioselective reductive opening of benzylidene acetal glycosides and polarity reversal.
Solubility
Decomposes with water.
Notes
Moisture Sensitive, store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Borane-trimethylamine complex | |
93°C to 95°C | |
172°C | |
C3H12BN | |
25 g | |
3905060 | |
WVMHLYQJPRXKLC-UHFFFAOYSA-N | |
trimethylamine borane | |
72.95 |
75-22-9 | |
0.81 | |
64°C (147°F) | |
MFCD00012420 | |
UN1325 | |
Decomposes with water. | |
B.CN(C)C | |
72.95 | |
97% |
Chemical Identifiers
75-22-9 | |
72.95 | |
WVMHLYQJPRXKLC-UHFFFAOYSA-N | |
B.CN(C)C |
C3H12BN | |
MFCD00012420 | |
trimethylamine borane |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P374
H228-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-850-4
RTECSNumber : PA0525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only