Please login to your online account to display your discounted pricing

Borane-triethylamine Complex 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC402731000

 View more versions of this product

Catalog No. AC402731000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0% (based on active hydrogen)
Physical Form Liquid
Formula Weight 115.03
Quantity 100g
Color Clear colorless
Beilstein 04, IV, 329
EINECS Number 217-022-3
TSCA TSCA
Fieser 03,299; 01,1229; 05,708; 12,65
Packaging Glass bottle
Name Note 99%
Assay Percent Range 97%
Linear Formula (C2H5)3N·BH3
MDL Number MFCD00012423
Synonym TEAB
CAS 1722-26-5
Chemical Name or Material Borane-triethylamine complex
Molecular Formula C6H18BN
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material Borane-triethylamine complex
Name Note 99%
CAS 1722-26-5
Assay ≥96.0% (based on active hydrogen)
Assay Percent Range 97%
Molecular Formula C6H18BN
Linear Formula (C2H5)3N·BH3
Formula Weight 115.03
MDL Number MFCD00012423
Synonym TEAB
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

SDS & Safety Documents