Please login to your online account to display your discounted pricing

Borane-triethylamine Complex 99%, ACROS Organics™


Quantity 25g
Packaging Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC402730250

 View more versions of this product

Catalog No. AC402730250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (based on active hydrogen)
Formula Weight 115.03
Physical Form Liquid
Quantity 25g
Color Clear colorless
Packaging Glass Bottle in Tin Can
EINECS Number 217-022-3
TSCA TSCA
Density 0.77g/mL
Fieser 03,299; 01,1229; 05,708; 12,65
Beilstein 04, IV, 329
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Linear Formula (C2H5)3N·BH3
MDL Number MFCD00012423
Name Note 99%
Molecular Formula C6H18BN
Assay Percent Range 97%
CAS 1722-26-5
Chemical Name or Material Borane-triethylamine complex
Synonym TEAB
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Borane-triethylamine complex
Name Note 99%
CAS 1722-26-5
Assay ≥96.0% (based on active hydrogen)
Assay Percent Range 97%
Molecular Formula C6H18BN
Linear Formula (C2H5)3N·BH3
Formula Weight 115.03
MDL Number MFCD00012423
Synonym TEAB
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

SDS & Safety Documents