Please login to your online account to display your discounted pricing

Borane-triethylamine Complex 99%, ACROS Organics™

$250.64 - $250.64

Chemical Identifiers

Chemical Name or Material Borane-triethylamine complex
Name Note 99%
CAS 1722-26-5
Assay ≥96.0% (based on active hydrogen)
Assay Percent Range 97%
Molecular Formula C6H18BN
Linear Formula (C2H5)3N·BH3
Formula Weight 115.03
MDL Number MFCD00012423
Synonym TEAB
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC402730250

  SDS
thermo fisher scientific
AC402730250
Glass Bottle in Tin Can 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC402731000

  SDS
thermo fisher scientific
AC402731000
Glass bottle 100g Each for $250.64
Description & Specifications

Specifications

Assay ≥96.0% (based on active hydrogen)
Physical Form Liquid
Formula Weight 115.03
Quantity 25g
Color Clear colorless
Packaging Glass Bottle in Tin Can
EINECS Number 217-022-3
TSCA TSCA
Density 0.77g/mL
Beilstein 04, IV, 329
Fieser 03,299; 01,1229; 05,708; 12,65
Linear Formula (C2H5)3N·BH3
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Name Note 99%
Molecular Formula C6H18BN
MDL Number MFCD00012423
Assay Percent Range 97%
CAS 1722-26-5
Chemical Name or Material Borane-triethylamine complex
Synonym TEAB
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Reduction reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.