Please login to your online account to display your discounted pricing

Alfa Aesar™ Borane-triethylamine complex, 96%, packaged under Argon in resealable ChemSeal™ bottles

As a reducing agent and in hydroboration reactions

Chemical Identifiers

Chemical Name or Material Borane-triethylamine complex
CAS 1722-26-5
Molecular Formula C6H15N·BH3
Linear Formula (C2H5)3N·BH3
Formula Weight 115.03
MDL Number MFCD00012423
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4296214

  SDS
alfa aesar™
4296214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 115.03
Chemical Name or Material Borane-triethylamine complex
Packaging ChemSeal™ bottles
TSCA TSCA
Physical Form Liquid
Assay 96%
Molecular Formula C6H15N·BH3
Linear Formula (C2H5)3N·BH3
MDL Number MFCD00012423
CAS 1722-26-5
UN Number UN2924
Melting Point −4°C
Color Colorless to pale yellow
Boiling Point 97°C (12mmHg)
Quantity 25g
Flash Point −6°C (21°F)
Odor Fish-like
Sensitivity Moisture Sensitive
Density 0.777g/mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Packaged under Argon
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

  • Packaged under Argon