missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Borane-tetrahydrofuran complex, 1M solution in THF, Stabilized, AcroSeal™, ACROS Organics™

Manufacturer:  Acros Organics AC175081000

 View more versions of this product

Catalog No. AC175081000


 

Description

Description

Reduction reagent
Specifications

Specifications

Borane-tetrahydrofuran complex
14044-65-6,109-99-9
0.96 to 1.08M
0.8760g/mL
15, 1336
[B].C1CCOC1
82.917
85.94
AcroSeal™ Glass bottle
0.005M sodium borohydride max.
Stabilized
100mL
1M solution in tetrahydrofuran, stabilized
C4H8BO
MFCD00012429
01,199; 02,106; 03,76; 04,124; 05,184; 06,161; 07,89; 12,65; 17,101
UWTDFICHZKXYAC-UHFFFAOYSA-N
boron;oxolane
11062302
Liquid
Solubility in water: reacts.
0.876
Colorless
−22°C

Chemical Identifiers

14044-65-6
82.917
UWTDFICHZKXYAC-UHFFFAOYSA-N
C4H8BO
MFCD00012429
11062302
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation. Causes serious eye damage. In contact with water releases flammable gases which may ignite spontaneously. Causes skin irritation. Harmful if swallowed. Highly flammable liquid and vapour. Suspected of causing cancer. Reacts violently with water. May form explosive peroxides. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

EINECSNumber : 237-881-8

TSCA : TSCA

SDS Documents