Please login to your online account to display your discounted pricing

Alfa Aesar™ Borane-tetrahydrofuran complex, 1M soln. in THF, stab. with 5mmol NaBH4

$80.61 - $313.41

Chemical Identifiers

Chemical Name or Material Borane-tetrahydrofuran complex
CAS 14044-65-6
Name Note stabilized with 5mmol NaBH4
Molecular Formula C4H11BO
Linear Formula BH3/(CH2)4O
Formula Weight 85.94
MDL Number MFCD00012429
UN Number UN3148
Beilstein 3668402
Synonym BTHF
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL13961AE

  SDS
alfa aesar™
L13961AE
100mL Each for $80.61

AAL13961AP

  SDS
alfa aesar™
L13961AP
500mL Each for $313.41
Description & Specifications

Specifications

Formula Weight 85.94
Chemical Name or Material Borane-tetrahydrofuran Complex
Quantity 100mL
Concentration 1M soln. in THF
Beilstein 3668402
Linear Formula BH3/(CH2)4O
UN Number UN3148
Recommended Storage 2° to 8°C
CAS 14044-65-6
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point −21°C (−8°F)
Health Hazard 1 Danger
Synonym BTHF
Health Hazard 2 GHS H Statement
H225-H260-H318-H351-H302-H335-H315
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Molecular Formula C4H11BO
TSCA TSCA
Name Note stabilized with 5mmol NaBH4
DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: WATER-REACTIVE LIQUID, N.O.S.
MDL Number MFCD00012429

Applications
Borane-tetrahydrofuran complex is an important reagent used in the reduction of certain functional groups viz. aldehyde, ketone, carboxylic acid, amide, oxime, imine and nitrile. It is also used as hydro borating agent. It acts as a borane source for oxazaborolidine catalyzed asymmetric reductions. It is utilized to reduce nylon surface amide groups to secondary amines.

Solubility
Miscible and reacts violently with water.

Notes
Air and moisture sensitive. Forms explosive peroxides in contact with air. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and alcohols. On hydrolysis, it forms hydrogen and boric acid.

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H260-H318-H351-H302-H335-H315
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

2° to 8°C