Please login to your online account to display your discounted pricing

Alfa Aesar™ Borane-tetrahydrofuran complex, 1M soln. in THF, packaged under Argon in resealable ChemSeal™ bottles

$61.88 - $217.04

Chemical Identifiers

Chemical Name or Material Borane-tetrahydrofuran complex
CAS 14044-65-6
Linear Formula BH3/(CH2)4O
Formula Weight 85.94
MDL Number MFCD00012429
UN Number UN3148
Beilstein 3668402
Synonym BTHF
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43291AP

 
alfa aesar™
43291AP
500mL Each for $217.04

AA43291AE

 
alfa aesar™
43291AE
100mL Each for $61.88
Description & Specifications

Specifications

Formula Weight 85.94
Health Hazard 2 GHS H Statement
H225-H260-H318-H351-H302-H335-H315
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Packaging ChemSeal™ bottles
TSCA TSCA
Color Colorless
Quantity 500mL
DOT Information Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: WATER-REACTIVE LIQUID, N.O.S.
Chemical Name or Material Borane-tetrahydrofuran complex
Physical Form Liquid
Linear Formula BH3/(CH2)4O
MDL Number MFCD00012429
CAS 14044-65-6
UN Number UN3148
Beilstein 3668402
Concentration 1M soln. in THF
Synonym BTHF
Sensitivity Air & Moisture Sensitive
Density 0.878g/mL
Health Hazard 1 Danger
Storage Note 1 2° to 8°C

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H260-H318-H351-H302-H335-H315
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

2° to 8°C