missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Borane-tert-butylamine complex, 97%

Manufacturer:  Alfa Aesar™ L1317718

 View more versions of this product

Catalog No. AAL1317718



Description

Description

Applications
Borane-tert-butylamine complex is used in stereoselective reduction of a steroidal ketone and for a review of amine boranes as selective reducing and hydroborating agents. It is also used as strong co-reducing agent in the syntheses of copper (I) oxide nanoparticles, diphosphine-protected gold nanocluster and 4-acetoxycinnamyl alcohols.

Solubility
Soluble in water (27 g/L at 20°C) and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and acids.
Specifications

Specifications

Borane-tert-butylamine complex
50g
C4H11BN
UN2811
tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane
[B].CC(C)(C)N
83.949
86.98
96°C to 98°C
7337-45-3
MFCD00075635
4134905
PVYPHUYXKVVURH-UHFFFAOYSA-N
boron;2-methylpropan-2-amine
6364547
97%

Chemical Identifiers

7337-45-3
83.949
PVYPHUYXKVVURH-UHFFFAOYSA-N
6364547
[B].CC(C)(C)N
C4H11BN
MFCD00075635
tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane
boron;2-methylpropan-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : EO3900000

TSCA : TSCA

Documents