Please login to your online account to display your discounted pricing

Borane-tert-butylamine complex, 95%, powder, ACROS Organics™


Quantity 25g
Packaging Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  177300250

 View more versions of this product

Catalog No. AC177300250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94% (active hydrogen)
Formula Weight 86.97
Physical Form Fine Crystalline Powder
Quantity 25g
Color White to off-white
Packaging Glass Bottle in Tin Can
TSCA TSCA
RTECS Number EO3900000
Melting Point 96° to 101°C
EINECS Number 230-851-5
Fieser 11,87
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Causes serious eye irritation.
Toxic in contact with skin.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Flammable solid.
Molecular Formula C4H14BN
Linear Formula (CH3)3CNH2·BH3
Assay Percent Range 95%
CAS 7337-45-3
MDL Number MFCD00075635
Name Note 95%
Chemical Name or Material Borane-tert-butylamine complex
CAS 75-64-9
Synonym tert-Butylamineborane
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Borane-tert-butylamine complex
Name Note 95%
CAS 7337-45-3
CAS 75-64-9
Assay ≥94% (active hydrogen)
Assay Percent Range 95%
Molecular Formula C4H14BN
Linear Formula (CH3)3CNH2·BH3
Formula Weight 86.97
MDL Number MFCD00075635
Synonym tert-Butylamineborane
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Causes serious eye irritation.
Toxic in contact with skin.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Flammable solid.

SDS & Safety Documents