Please login to your online account to display your discounted pricing

Alfa Aesar™ Borane-pyridine complex, 95%

Chemical Identifiers

Chemical Name or Material Borane-pyridine complex
CAS 110-51-0
Molecular Formula C5H8BN
Formula Weight 92.93
MDL Number MFCD00012435
UN Number UN2810
Beilstein 3674913
Synonym Pyridine-borane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1317809

  SDS
alfa aesar™
L1317809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1317818

  SDS
alfa aesar™
L1317818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 92.93
Chemical Name or Material Borane-pyridine complex
TSCA TSCA
Assay 95%
Molecular Formula C5H8BN
MDL Number MFCD00012435
CAS 110-51-0
Boiling Point 65°C (1mmHg)
Flash Point 73°C (163°F)
Odor Unpleasant
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210a-P280h-P305+P351+P338-P309-P310
Keep away from heat. - No smoking.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
UN Number UN2810
Melting Point 9° to 11°C
Beilstein 3674913
Synonym Pyridine-borane
Quantity 10g
Health Hazard 2 GHS H Statement
H301-H310-H330-H227-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECS Number US3675000

Applications
Borane-pyridine complex is used for synthesis of α-alkyl-β-hydroxy esters and preparation of Igs conjugated with viral peptide epitopes. It is used as reducing agent for high performance liquid chromatography analysis of oligosaccharides. It is also used as modifier in decant oil affecting its carbonization to mesophase pitch.

Solubility
It hydrolyzes slowly in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.

Safety and Handling

Danger

GHS P Statement
P210a-P280h-P305+P351+P338-P309-P310
Keep away from heat. - No smoking.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H310-H330-H227-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C