Please login to your online account to display your discounted pricing

Borane-morpholine complex, 97%, ACROS Organics™

$106.96 - $106.96

Chemical Identifiers

Chemical Name or Material Borane-morpholine complex
Name Note 97%
CAS 4856-95-5
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H12BNO
Formula Weight 100.96
MDL Number MFCD00066736
Synonym Morpholineborane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC177290250

  SDS
thermo fisher scientific
177290250
Glass bottle 25g Each for $106.96
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 100.96
Quantity 25g
Color Colorless to white
TSCA TSCA
EINECS Number 225-450-7
Melting Point 95° to 99°C
RTECS Number QD8530000
Chemical Name or Material Borane-morpholine complex
Packaging Glass bottle
Name Note 97%
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C4H12BNO
MDL Number MFCD00066736
CAS 4856-95-5
Synonym Morpholineborane
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable solid.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Reduction reagent

Safety and Handling

GHS Signal Word: Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable solid.