Please login to your online account to display your discounted pricing

Alfa Aesar™ Borane-morpholine complex, 97%

Chemical Identifiers

Chemical Name or Material Borane-morpholine complex
CAS 4856-95-5
Molecular Formula C4H12BNO
Formula Weight 100.96
MDL Number MFCD00066736
UN Number UN2925
Synonym Morpholine borane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1511006

 
alfa aesar™
L1511006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1511014

 
alfa aesar™
L1511014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.96
Chemical Name or Material Borane-morpholine complex
Molecular Formula C4H12BNO
MDL Number MFCD00066736
CAS 4856-95-5
Synonym Morpholine borane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228-H301-H314-H318
Flammable solid.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 97%
UN Number UN2925
Melting Point 93° to 95°C
Quantity 5g
Odor Amine-like
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
RTECS Number QD8530000
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H301-H314-H318
Flammable solid.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.