Learn More
Borane dimethyl sulfide complex, 94%, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 177061000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Borane dimethyl sulfide complex | |
13292-87-0,75-18-3 | |
Amber to Colorless | |
3°C | |
AcroSeal™ Glass bottle | |
(CH3)2S·BH3 | |
100 mL | |
0.801 | |
dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms | |
RMHDLBZYPISZOI-UHFFFAOYSA-N | |
boron;methylsulfanylmethane | |
9833925 | |
94% |
94% | |
-40°C to 37°C | |
0.8010g/mL | |
94% | |
C2H9BS | |
MFCD00013189 | |
04,191; 05,47; 06,64; 08,49; 10,49; 14,52; 17,50 | |
15, 1336 | |
Solubility in water: reacts. Other solubilities: soluble in ethyl ether,tetrahydrofurane,hexane,,toluene,methylene chloride and other aprotic,solvents | |
B.CSC | |
75.96 | |
75.95 | |
Solution |
Chemical Identifiers
13292-87-0 | |
75.96 | |
RMHDLBZYPISZOI-UHFFFAOYSA-N | |
9833925 | |
B.CSC |
C2H9BS | |
MFCD00013189 | |
dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms | |
boron;methylsulfanylmethane |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
In contact with water releases flammable gases which may ignite spontaneously.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
Harmful to aqua
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
GHS Signal Word: Danger
EINECSNumber : 236-313-6
RTECSNumber : PV5080000
TSCA : TSCA
RUO – Research Use Only