missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Borane-diphenylphosphine Complex, 94%, Thermo Scientific Chemicals
$300.57 - $300.57
Chemical Identifiers
CAS | 41593-58-2 |
---|---|
Molecular Formula | C12H14BP |
Molecular Weight (g/mol) | 200.03 |
MDL Number | MFCD00274254 |
InChI Key | XUFGOLYVDOBYLK-UHFFFAOYSA-N |
Synonym | borane-diphenylphosphine complex, diphenylphosphine borane, borane diphenylphosphine complex, diphenylphosphine-borane, diphenylphosphine trihydroboron, xufgolyvdobylk-uhfffaoysa-n, diphenylphosphine boranylidyne |
PubChem CID | 11095625 |
IUPAC Name | boron;diphenylphosphane |
SMILES | B.P(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC380750010
|
Thermo Scientific Chemicals
380750010 |
1 g | Glass bottle |
Each for $300.57
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
41593-58-2 | |
200.03 | |
XUFGOLYVDOBYLK-UHFFFAOYSA-N | |
11095625 | |
B.P(C1=CC=CC=C1)C1=CC=CC=C1 |
C12H14BP | |
MFCD00274254 | |
borane-diphenylphosphine complex, diphenylphosphine borane, borane diphenylphosphine complex, diphenylphosphine-borane, diphenylphosphine trihydroboron, xufgolyvdobylk-uhfffaoysa-n, diphenylphosphine boranylidyne | |
boron;diphenylphosphane |
Specifications
41593-58-2 | |
White | |
148.0°C to 150.0°C (10.0 mmHg) | |
94% | |
C12H14BP | |
1 g | |
Solubility in water: decomposes | |
B.P(C1=CC=CC=C1)C1=CC=CC=C1 | |
200.03 | |
200.03 | |
Crystals or Powder |
47.0°C to 50.0°C | |
92.0% min. ex C% min. | |
Authentic | |
Glass bottle | |
MFCD00274254 | |
borane-diphenylphosphine complex, diphenylphosphine borane, borane diphenylphosphine complex, diphenylphosphine-borane, diphenylphosphine trihydroboron, xufgolyvdobylk-uhfffaoysa-n, diphenylphosphine boranylidyne | |
XUFGOLYVDOBYLK-UHFFFAOYSA-N | |
boron;diphenylphosphane | |
11095625 | |
94% | |
Borane-diphenylphosphine complex |
Safety and Handling
GHS H Statement
In contact with water releases flammable gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas.
Protect from moisture.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.
GHS Signal Word: Danger
RUO – Research Use Only