missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Borane-dimethylamine complex, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 177311000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Reduction reagentSpecifications
Borane-dimethylamine complex | |
65°C | |
98+% | |
C2H10BN | |
100g | |
Solubility in water: 125g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,thf,,diglyme,benzene,toluene,chlorobenzene | |
58.92 |
74-94-2 | |
Authentic | |
Glass bottle | |
(CH3)2NH·BH3 | |
Dimethylamineborane, DMAB | |
58.92 | |
98+% |
Chemical Identifiers
74-94-2 | |
C2H10BN | |
58.92 | |
Dimethylamineborane, DMAB |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 200-823-7
RUO â Research Use Only