Please login to your online account to display your discounted pricing

Alfa Aesar™ Borane-dimethyl sulfide complex, nominally 5M in ether, packaged under Argon in resealable ChemSeal™ bottles

$298.62 - $298.62

Chemical Identifiers

Chemical Name or Material Borane-dimethyl sulfide complex
CAS 13292-87-0
Molecular Formula C2H9BS
Linear Formula (CH3)2S·BH3
Formula Weight 75.97
MDL Number MFCD00013189
UN Number UN2924
Beilstein 3663489
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43327AK

 
alfa aesar™
43327AK
250mL Each for $298.62
Description & Specifications

Specifications

Formula Weight 75.97
Health Hazard 2 GHS H Statement
H224-H261-H314-H318-H302-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P280-P243-P305+P351+P338-P370+P378i-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.
Chemical Name or Material Borane-dimethyl sulfide complex
Packaging ChemSeal™ bottles
Physical Form Liquid
Molecular Formula C2H9BS
Linear Formula (CH3)2S·BH3
CAS 13292-87-0
UN Number UN2924
Beilstein 3663489
Color Colorless
Quantity 250mL
Flash Point −35°C (−31°F)
Odor Characteristic
Sensitivity Air & Moisture Sensitive
Storage Note 1
  • Packaged under Argon
Recommended Storage 2°C to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA TSCA
MDL Number MFCD00013189
Concentration Nominally 5M in ether
Density 0.789g/mL
Health Hazard 1 Danger

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P280-P243-P305+P351+P338-P370+P378i-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

GHS H Statement
H224-H261-H314-H318-H302-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.

2°C to 8°C

  • Packaged under Argon