Learn More
Borane-dimethyl sulfide complex, 2M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 042965AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Borane-dimethyl sulfide complex | |
Colorless to Yellow | |
−17°C (−1°F) | |
ChemSeal™ bottles | |
(CH3)2S·BH3 | |
100 mL | |
3663489 | |
dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms | |
RMHDLBZYPISZOI-UHFFFAOYSA-N | |
boron;methylsulfanylmethane | |
9833925 | |
2M in THF |
13292-87-0 | |
0.855 g/mL | |
Characteristic | |
C2H9BS | |
MFCD00013189 | |
UN3399 | |
Air and moisture sensitive | |
Miscible with ethyl ether,dichloromethane,benzene,xylene,hexane,diglyme,dimethyl ether,ethyl acetate,toluene,methylene chloride and other aprotic solvents. | |
B.CSC | |
75.96 | |
75.97 | |
Liquid |
Chemical Identifiers
13292-87-0 | |
75.96 | |
RMHDLBZYPISZOI-UHFFFAOYSA-N | |
9833925 | |
B.CSC |
C2H9BS | |
MFCD00013189 | |
dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms | |
boron;methylsulfanylmethane |
Safety and Handling
GHS H Statement
H225-H261-H318-H302-H335-H315
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P335+P334
H225-H260-H302-H315-H318-H335-H336-H351
DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA : Yes
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only