Please login to your online account to display your discounted pricing

Alfa Aesar™ Borane-di-tert-butylphosphine complex, 98%

Chemical Identifiers

Chemical Name or Material Borane-di-tert-butylphosphine complex
CAS 128363-76-8
Molecular Formula C8H22BP
Linear Formula [(CH3)3C]2PH·BH3
Formula Weight 160.05
MDL Number MFCD03412071
UN Number UN2813
Synonym Di-tert-butylphosphine-borane complex
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2689903

 
alfa aesar™
H2689903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2689906

 
alfa aesar™
H2689906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.05
Chemical Name or Material Borane-di-tert-butylphosphine complex
Assay 98%
Molecular Formula C8H22BP
Linear Formula [(CH3)3C]2PH·BH3
MDL Number MFCD03412071
CAS 128363-76-8
UN Number UN2813
Synonym Di-tert-butylphosphine-borane complex
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P231+P232-P261-P280-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P231+P232-P261-P280-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.