Please login to your online account to display your discounted pricing

Borane-di(tert-butyl)phosphine complex, 94%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC380730010

 View more versions of this product

Catalog No. AC380730010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Needles
Formula Weight 160.05
Quantity 1g
Color White
Packaging Glass bottle
CAS 128363-76-8
Chemical Name or Material Borane-di(tert-butyl)phosphine complex
Assay Percent Range 94%
Molecular Formula C8H22BP
MDL Number MFCD03412071
Health Hazard 1 GHS Signal Word: Danger
Elemental Analysis 92.0% min.
Health Hazard 2 GHS H Statement:
In contact with water releases flammable gas.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Borane-di(tert-butyl)phosphine complex
Assay Percent Range 94%
Molecular Formula C8H22BP
Formula Weight 160.05
MDL Number MFCD03412071
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
In contact with water releases flammable gas.

SDS & Safety Documents