missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Borane-d3, 1M in tetrahydrofuran, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 44188AB

 View more versions of this product

Catalog No. AA44188AB



Description

Description

  • Packaged under Argon
Specifications

Specifications

Borane-d_3
ChemSeal™ bottles
Colorless
13763-62-7
BD3/(CH2)4O
UN3148
1M in tetrahydrofuran
B
16.852
16.84 as BD3
1M in tetrahydrofuran
-21°C (-6°F)
0.876g/mL
Air & Moisture Sensitive
10mL
BH3
MFCD08705283
borane-d3
UORVGPXVDQYIDP-FIBGUPNXSA-N
trideuterioborane
11062299
Liquid
Ether-like

Chemical Identifiers

13763-62-7
16.852
UORVGPXVDQYIDP-FIBGUPNXSA-N
11062299
B
BH3
MFCD08705283
borane-d3
trideuterioborane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H260-H318-H351-H302-H335-H315
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H225-H260-H302-H314-H335-H336-H351

DOTInformation : Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: WATER-REACTIVE LIQUID, N.O.S.

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Argon

Documents