Please login to your online account to display your discounted pricing

Alfa Aesar™ Borane-d3, 1M in tetrahydrofuran, packaged under Argon in resealable ChemSeal™ bottles

$643.39 - $643.39

Chemical Identifiers

Chemical Name or Material Borane-d_3
CAS 13763-62-7
Linear Formula BD3/(CH2)4O
Formula Weight 16.84 as BD3
MDL Number MFCD08705283
UN Number UN3148
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44188AD

  SDS
alfa aesar™
44188AD
50mL Each for $643.39

AA44188AB

  SDS
alfa aesar™
44188AB
10mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 16.84 as BD3
Health Hazard 2 GHS H Statement
H225-H260-H318-H351-H302-H335-H315
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 50mL
Physical Form Liquid
Density 0.876g/mL
Linear Formula BD3/(CH2)4O
UN Number UN3148
Chemical Name or Material Borane-d_3
MDL Number MFCD08705283
Concentration 1M in tetrahydrofuran
CAS 13763-62-7
Health Hazard 1 Danger
Color Colorless
Packaging ChemSeal™ bottles
Sensitivity Air & Moisture Sensitive
Storage Note 1 2° to 8°C
Odor Ether-like
DOT Information Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: WATER-REACTIVE LIQUID, N.O.S.

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H260-H318-H351-H302-H335-H315
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

2° to 8°C