missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Borane-d3, 1M in tetrahydrofuran

Manufacturer:  Alfa Aesar™ 89121AD

 View more versions of this product

Catalog No. AA89121AD



Specifications

Specifications

Borane-d_3
Ether-like
Colorless
13763-62-7
MFCD08705283
borane-d3, borane-d3, 1m in tetrahydrofuran, borane-d3, 1m in tetrahydrofuran, packaged under argon in resealable chemseal bottles
UORVGPXVDQYIDP-FIBGUPNXSA-N
trideuterioborane
11062299
Liquid
0.876g/mL
Air & Moisture Sensitive
50mL
BH3
UN3148
1M in tetrahydrofuran
B
16.852
16.84 as BD3

Chemical Identifiers

13763-62-7
16.852
UORVGPXVDQYIDP-FIBGUPNXSA-N
11062299
B
BH3
MFCD08705283
borane-d3, borane-d3, 1m in tetrahydrofuran, borane-d3, 1m in tetrahydrofuran, packaged under argon in resealable chemseal bottles
trideuterioborane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H260-H318-H351-H302-H335-H315
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: WATER-REACTIVE LIQUID, N.O.S.

storageNote1 : 2° to 8°C

SDS Documents